Iridium ≥99,95% (Metall-Basis), Pulver -60 mesh

Lieferant: Thermo Fisher Scientific

12070.02 12070.04 12070.09
12070.02EA 408 EUR
12070.02 12070.04 12070.09
Iridium ≥99,95% (Metall-Basis), Pulver -60 mesh
Iridium

Purity: 99.95% CAS No.: 7439-88-5 Appearance: Grey MDL No.: MFCD00011062

Formel: Ir
Siedepunkt: 4500 °C (1013 hPa)
Schmelzpunkt: 2443 °C
Dichte: 22,65 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011062
CAS-Nummer: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III
Merck Index: 12,05098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR