Iridium, schwarz ≥99,8% (Metall-Basis), Pulver

Lieferant: Thermo Fisher Scientific
Achtung

12071.02 12071.04 12071.09
12071.02EA 408 EUR
12071.02 12071.04 12071.09
Iridium, schwarz ≥99,8% (Metall-Basis), Pulver
Iridium

Purity: 99.8% CAS No.: 7439-88-5 Physical Form: S.A. 20m2/g UN No.: 3089 MDL No.: MFCD00011062

Formel: Ir
Lagertemperatur: Raumtemperatur
MDL: MFCD00011062
CAS-Nummer: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III
Merck Index: 12,05098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR