Absorptions-Saugstrümpfe, PIG® Oil-Only
Lieferant: NEW PIG
SKM210
121-0128EA 81 EUR
121-0128
Absorptions-Saugstrümpfe, PIG® Oil-Only
Absorptionsmaterial
PIG® Oil-Only Absorbent Socks absorbs and retains oils and oil-based liquids - including lubricants and fuels - without absorbing a drop of water.
  • Outlasts other outdoor socks with the most UV resistant construction
  • Moldable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and keep workers safe
  • Cellulose filler is hydrophobic for containing and absorbing oil-based spills in a variety of environments
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Ideal for use on land, but also floats to clean up oil-based liquids on water
  • Attached string conveniently allows socks to be tied down, tied together or dipped in non-corrosive wash baths
  • Skin is UV resistant up to 12 months; meets NFPA 99 standards for static decay
95% mixture of pre- and post-consumer recycled cellulose. Not recommended for use with acids, caustics or other corrosives.