PIG® eco-friendly compact spill kit
Lieferant: NEW PIG
KITE210
121-0191EA 22 EUR
121-0191
PIG® eco-friendly compact spill kit
Absorptionsmaterial
This compact eco-friendly kit makes transport simple, and its recycled content pads and pillow soak up more liquid with less environmental impact.
  • Environmentally friendly absorbents feature high recycled content; reuses material that would normally enter a waste stream
  • Reusable wallet is constructed out of jute plant bark; naturally strong and durable with waterproof inner coating to keep the kit clean, dry and ready to use
  • High visibility colour helps identify spill kit location; saves valuable time during response
  • ‘Hook and loop’ closure is easy to open for fast access to absorbents
  • Carrying handle affords convenient transport; makes it ideal for hanging on a wall, storing in a cabinet or being carried in a vehicle cab
  • Compact design takes up little storage room and effectively deals with minor spillages
Super absorbent PIG BLUE® mat pads and PIG® pillow have maximum capacity for soaking up spills of oils, coolants, solvents or water.