VWR®, Chemisches Absorptionskit, 16 L
Lieferant: VWR Collection
121-0194EA 88.3 EUR
121-0194
VWR®, Chemisches Absorptionskit, 16 L
Absorptionsmaterial
This portable, economy spill kit comes in a high visibility, water-resistant, yellow PVC bag with handles. Designed for small spills of chemicals and hazardous liquids. Ideal for small storage areas, thanks to its compact size.
  • Fast spill response
  • Yellow colour for high visibility
  • Water-resistant, lightweight bag keeps sorbents dry
  • Can fit behind or under a seat in a vehicle
All you need for an emergency spill response in a compact and high vis carry bag. Ideal for workers on the go, like field workers, or for carriers and truck drivers who occasionally transport small quantities of liquids.
Lieferumfang: Jedes Kit enthält 10 VWR®-Pads (41×51 cm), 2 VWR®-Schlangen (Ø 7,6×122 cm), 1 Paar Handschuhe, 1 Entsorgungsbeutel und einen 10-Stufen-Plan, der auch das Vorgehen bei einem Verschütten beschreibt.
Verpackung: In einem haltbaren, gelben PVC-Beutel mit Griffen verpackt.