Silber ≥99,9985% (Metall-Basis), Folie, Premion®, Dicke 0.5 mm (0.02 in)
Lieferant: Thermo Fisher Scientific
12128.FF
12128.FI
12128.GH
12128.FFEA
71.4
EUR
12128.FF
12128.FI
12128.GH
Silber ≥99,9985% (Metall-Basis), Folie, Premion®, Dicke 0.5 mm (0.02 in)
Silver
Purity: 99.9985%
CAS No.: 7440-22-4
Physical Form: ≈3.3g/25x25mm
Odour: Characteristic
MDL No.: MFCD00003397
Formel:
Ag Siedepunkt: 2212 °C (1013 hPa) Schmelzpunkt: 962 °C Dichte: 10,5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003397 CAS-Nummer: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...