Bor ≥99,5% (Metall-Basis), kristalline Stücke ≤ 20 mm (0.8 in)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12134.09
12134.18
12134.30
12134.09EA
156
EUR
12134.09
12134.18
12134.30
Bor ≥99,5% (Metall-Basis), kristalline Stücke ≤ 20 mm (0.8 in)
Boron
Purity: 99.5%
CAS No.: 7440-42-8
Odour: Odourless
MDL No.: MFCD00134034
Formel:
B Dichte: 1,73 g/cm³ (15 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134034 CAS-Nummer: 7440-42-8 EINECS: 231-151-2 Merck Index: 13,01333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...