Rhodanile blue 75% (Farbstoffgehalt), fest, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ100

121495-5G
121495-5GEA 186 EUR
121495-5G
Rhodanile blue 75% (Farbstoffgehalt), fest, Sigma-Aldrich®
Rhodanile blue
Formel: C₄₈H₄₈ClN₅O₃
Molecular Weight: 778.40 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011937
CAS-Nummer: 14969-56-3
EINECS: 239-040-0

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Very Dark Purple, Blue or Black
Appearance (form) Powder or crystals
Infrared spectrum Conforms to structure
Carbon 53.9 - 90.8 %
Nitrogen 6.5 - 11.0 %
Wavelength in water 634 - 640 nm
Extinction coefficient ≥90000
Wavelength 552 - 558 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR