Chromdisilicid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chromium silicide

12151.18 12151.30
12151.18EA 66 EUR
12151.18 12151.30
Chromdisilicid ≥99% (Metall-Basis)
Chromium disilicide

Purity: 99+% CAS No.: 12018-09-6 Molecular Formula: CrSi2 Formula Weight: 108.17 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00168070

Formel: CrSi₂
Molecular Weight: 108.17 g/mol
Schmelzpunkt: 1490 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00168070
CAS-Nummer: 12018-09-6
EINECS: 234-633-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR