2-Aminoazotoluol ≥97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
121568-100G
121568-100GEA
151
EUR
121568-100G
2-Aminoazotoluol ≥97%, Sigma-Aldrich®
2-Aminoazotoluol
Fast Garnet GBC base has been used as a stain in papain activity inhibition assay, to stain for tryptase-activity and chymase-like activity, to gel stain for esterolytic activity.
Formel:
C₁₄H₁₅N₃ Molecular Weight: 225,29 g/mol Schmelzpunkt: 101,..102 °C (Lit.) |
MDL:
MFCD00007733 CAS-Nummer: 97-56-3 EINECS: 202-591-2 |
Spezifikation Testergebnisse
Appearance (Color) | Orange to Dark Brown |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Wavelength | 325 - 331 nm |
Extinction Coefficient | ≥2000 |
Extinction Coefficient 0.01g/l, 50% Ethanol in 1N HCl | ≥17500 |
Carbon | ≥72.0 % |
Nitrogen | ≥17.9 % |
Purity (TLC) | ≥97 % |
Solubility (Turbidity) | Clear |
Solubility (Color), 1mg/ml, Ethanol | Orange to Dark Brown |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...