Gold(III)bromid ≥99,99% (Metall-Basis) Au ≥ 44.6%, Premion®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Goldtribromid
12162.03
12162.03EA
142
EUR
12162.03
Gold(III)bromid ≥99,99% (Metall-Basis) Au ≥ 44.6%, Premion®
Gold(III)bromid
Purity: 99.99%
CAS No.: 10294-28-7
Molecular Formula: AuBr3
Formula Weight: 436.69
Physical Form: Granular
Melting Point: ca 160° dec.
UN No.: 3260
MDL No.: MFCD00014171
Formel:
AuBr₃ Molecular Weight: 436.68 g/mol Schmelzpunkt: ∼ 160 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014171 CAS-Nummer: 10294-28-7 EINECS: 233-654-2 UN: 3260 ADR: 8 III Merck Index: 12,04539 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...