Gold(III)chlorid (≥64,4% Au)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Goldtrichlorid

12163.01 12163.03 12163.06
12163.01EA 93 EUR
12163.01 12163.03 12163.06
Gold(III)chlorid (≥64,4% Au)
Gold(III)chlorid

CAS No.: 13453-07-1 Molecular Formula: AuCl3 Formula Weight: 303.33 Physical Form: Crystalline Density: 3.90 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: 180(subl) Merck Reference: 14,4521 UN No.: 3260 MDL No.: MFCD00014172

Formel: AuCl₃
Molecular Weight: 303.32 g/mol
Schmelzpunkt: 180 °C (Subl.)
Dichte: 3.90
Lagertemperatur: Raumtemperatur
MDL: MFCD00014172
CAS-Nummer: 13453-07-1
EINECS: 236-623-1
UN: 3260
ADR: 8 III
Merck Index: 13,04536

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR