Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99,95% (Metall-Basis), Premion®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Disodium tetrachloroplatinate hydrate, Sodium platinum chloride hydrate, Sodium chloroplatinate(II) hydrate

12168.03 12168.06
12168.03EA 145 EUR
12168.03 12168.06
Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99,95% (Metall-Basis), Premion®
Sodium tetrachloroplatinate(II) hydrate

Purity: 99.95% CAS No.: 207683-21-4 Molecular Formula: Na2PtCl4.xH2O (x≈3) Formula Weight: 382.88(anhy) Physical Form: Crystalline Melting Point: 100° MDL No.: MFCD00150188

Formel: Na₂PtCl₄·xH₂O
Schmelzpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150188
CAS-Nummer: 207683-21-4
EINECS: 233-051-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR