di-Kaliumhexachloroplatinat (39,6% Pt)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 16921-30-5
Molecular Formula: Cl6K2Pt
Formula Weight: 486.01
Physical Form: Crystalline powder
Density: 3.50
Refractive Index: ≈1.825
Sensitivity: Hygroscopic
Appearance: Yellow
Odour: Odourless
Melting Point: 250°dec.
Solubility: Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol
Merck Reference: 14,7636
UN No.: 3288
MDL No.: MFCD00011389
Formel:
K₂PtCl₆ Schmelzpunkt: 250 °C, (Zersetzung) Dichte: 3,50 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011389 CAS-Nummer: 16921-30-5 EINECS: 240-979-3 UN: 3288 ADR: 6.1,III Merck Index: 14,7636 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...