Platin(IV)bromid (≥37,1% Pt) ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
12171.03
12171.03EA
102
EUR
12171.03
Platin(IV)bromid (≥37,1% Pt) ≥99,99% (Metall-Basis)
Platin(IV)bromid
Purity: 99.99%
CAS No.: 68938-92-1
Molecular Formula: Br4Pt
Formula Weight: 514.73
Physical Form: Crystalline
Appearance: Brown to black
Melting Point: 180° dec.
UN No.: 3260
MDL No.: MFCD00016289
Formel:
Br₄Pt Molecular Weight: 514,7 g/mol Schmelzpunkt: 180 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016289 CAS-Nummer: 68938-92-1 EINECS: 273-151-5 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...