Iridium(IV)chlorid (≥56,5% Ir) ≥99,95% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12184.06
12184.06EA
2130
EUR
12184.06
Iridium(IV)chlorid (≥56,5% Ir) ≥99,95% (Metall-Basis), Premion®
Iridium(IV)chlorid
Purity: 99.95%
CAS No.: 10025-97-5
Molecular Formula: IrCl4
Formula Weight: 334.01
Physical Form: Glassy amorphous pieces/powder
Sensitivity: Hygroscopic
Appearance: Dark brown to black
Melting Point: dec.
MDL No.: MFCD00016155
Formel:
Cl₄Ir Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016155 CAS-Nummer: 10025-97-5 EINECS: 233-048-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...