Agarose, Millipore®
Lieferant: Merck Millipore Calbiochem
121852-100
121852-100EA
543
EUR
121852-100
Agarose, Millipore®
Agarose
Schmelzpunkt:
60…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081294 CAS-Nummer: 9012-36-6 EINECS: 232-731-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...