Indium ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)
Lieferant: Thermo Fisher Scientific
12206.FI
12206.FY
12206.GJ
12206.HW
12206.FIEA
95.7
EUR
12206.FI
12206.FY
12206.GJ
12206.HW
Indium ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)
Indium
Purity: 99.99%
CAS No.: 7440-74-6
Physical Form: ≈2.3g/50x50mm
Odour: Odourless
MDL No.: MFCD00134048
Formel:
In Siedepunkt: 2000 °C (1013 hPa) Schmelzpunkt: 156 °C Dichte: 7,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134048 CAS-Nummer: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...