Cadmium(II)carbonat ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
12229.30
12229.30EA
101
EUR
12229.30
Cadmium(II)carbonat ≥99% (Metall-Basis)
Cadmium(II)carbonat
Purity: 99+%
CAS No.: 513-78-0
Molecular Formula: CdCO3
Formula Weight: 172.42
Physical Form: Powder
Density: 4.26
Melting Point: 500° dec.
Solubility: Very slightly soluble in water. Soluble in dilute acids, concentrated solutions of ammonium salts
Merck Reference: 14,1616
UN No.: 2570
MDL No.: MFCD00010918
Formel:
CCdO₃ Molecular Weight: 172,42 g/mol Schmelzpunkt: 500 °C (Zers.) Dichte: 4,26 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010918 CAS-Nummer: 513-78-0 EINECS: 208-168-9 UN: 2570 ADR: 6.1,III Merck Index: 14,1616 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...