Bismut(III)oxid ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: Powder
Density: 8.9
Appearance: Yellow
Melting Point: 817°
Boiling Point: ca 1890°
Solubility: Practically insoluble in water. Soluble in acids
Merck Reference: 14,1273
MDL No.: MFCD00003462
In disinfectants, magnets, glass, rubber, vulcanization, in fireproofing papers and polymers, and in catalysts
Formel:
BiO₃ Siedepunkt: ∼1890 °C Schmelzpunkt: 817 °C Dichte: 8,9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003462 CAS-Nummer: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...