Quecksilber(II)chlorid ≥99,5% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5% min
CAS No.: 7487-94-7
Molecular Formula: Cl2Hg
Formula Weight: 271.50
Physical Form: Powder
Density: 5.4
Refractive Index: 1.859
Appearance: White
Odour: Odourless
Melting Point: 280-282°
Boiling Point: 302°
Solubility: Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides), alcohol, glycerol, methanol, acetone, and ethyl acetate
Merck Reference: 14,5876
UN No.: 1624
MDL No.: MFCD00011041
Formel:
HgCl₂ Siedepunkt: 302 °C Schmelzpunkt: 280…282 °C Dichte: 5,4 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011041 CAS-Nummer: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1,II Merck Index: 14,5876 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...