di-Bortrioxid ≥97,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12290.36
12290.A3
12290.A9
12290.36EA
40.5
EUR
12290.36
12290.A3
12290.A9
di-Bortrioxid ≥97,5%
di-Bortrioxid
Purity: 97.5% min
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Physical Form: -60 Mesh
Density: 2.46 +/- 0.01
Appearance: White
Odour: Odourless
Melting Point: ca 450°
Boiling Point: 1860°
Solubility: Slightly soluble in water @ 25°C. Soluble in water @ 100°C
Merck Reference: 14,1337
MDL No.: MFCD00011315
As a flux, acid catalyst, or chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides
Formel:
B₂O₃ Siedepunkt: 1860 °C Schmelzpunkt: ∼450 °C Dichte: 2,46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011315 CAS-Nummer: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...