Cobalt(II)chlorid, wasserfrei ≥99,7% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.7%
CAS No.: 7646-79-9
Molecular Formula: Cl2Co
Formula Weight: 129.84
Physical Form: Powder
Density: 3.367
Appearance: Pale blue
Odour: Odourless
Melting Point: 735°
Boiling Point: 1049°
Solubility: Soluble in water, alcohol, acetone, ether, glycerol, and pyridine
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00010938
Formel:
Cl₂Co Siedepunkt: 1049 °C Schmelzpunkt: 735 °C Dichte: 3,367 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010938 CAS-Nummer: 7646-79-9 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...