Tetrachlorgold(III)säure Hydrat (≥49% Au) ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tetrachlorgoldsäure Hydrat, Hydrogentetrachloroaurat(III) Hydrat

12325.03 12325.06 12325.14 12325.22 AB202976.0001 AB202976.0005
12325.03EA 189 EUR
12325.03 12325.06 12325.14 12325.22 ABCRAB202976.0001 ABCRAB202976.0005
Tetrachlorgold(III)säure Hydrat (≥49% Au) ≥99,9% (Metall-Basis)
Tetrachlorgold(III)säure Hydrat

Purity: 99.9% CAS No.: 27988-77-8 Molecular Formula: HAuCl4.xH2O Formula Weight: 339.79(anhy) Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Yellow-orange UN No.: 3260 MDL No.: MFCD00149903

Formel: HAuCl₄·H₂O
Molecular Weight: 357.8 g/mol
Dichte: 1.169 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149903
CAS-Nummer: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR