Chrom(III)chlorid, wasserfrei 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chromium trichloride

12336.18 12336.30
12336.18EA 90.4 EUR
12336.18 12336.30
Chrom(III)chlorid, wasserfrei 98%
Chrom(III)chlorid

Purity: 98% CAS No.: 10025-73-7 Molecular Formula: CrCl3 Formula Weight: 158.35 Physical Form: Crystalline Density: 2.87 Appearance: Violet Boiling Point: 1300(subl) Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00010948

Formel: Cl₃Cr
Molecular Weight: 158.35 g/mol
Siedepunkt: 1300 °C (Subl.)
Schmelzpunkt: 1152
Dichte: 2.87
Lagertemperatur: Raumtemperatur
MDL: MFCD00010948
CAS-Nummer: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 14,2222

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR