Calcium zirconium oxide ≥99,2% (Metall-Basis)
Lieferant: Thermo Scientific
12344.A1
12344.30
12344.A1EA
86.3
EUR
12344.A1
12344.30
Calcium zirconium oxide ≥99,2% (Metall-Basis)
Calcium zirconium oxide
Purity: 99.2%
CAS No.: 12013-47-7
Molecular Formula: CaZrO3
Formula Weight: 179.30
Physical Form: -325 Mesh Powder
Odour: Odourless
Melting Point: 2550°
MDL No.: MFCD00015982
Formel:
CaZrO₃ Molecular Weight: 179,30 g/mol Schmelzpunkt: 2550 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015982 CAS-Nummer: 12013-47-7 EINECS: 234-586-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...