Nickel(II)oxid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

12359.18 12359.30
12359.18EA 57.5 EUR
12359.18 12359.30
Nickel(II)oxid ≥99% (Metall-Basis)
Nickel(II)oxid

Purity: 99% CAS No.: 1313-99-1 Molecular Formula: NiO Formula Weight: 74.71 Physical Form: -325 Mesh Powder Density: 6.67 Melting Point: 1984° Merck Reference: 14,6512 MDL No.: MFCD00011145

Formel: NiO
Schmelzpunkt: 1984 °C
Dichte: 6,67
Lagertemperatur: Raumtemperatur
MDL: MFCD00011145
CAS-Nummer: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR