Aluminiumhydroxid ≥76,5%

Lieferant: Thermo Fisher Scientific
Achtung

12366.22 12366.36 12366.A3
12366.22EA 29.2 EUR
12366.22 12366.36 12366.A3
Aluminiumhydroxid ≥76,5%
Aluminiumhydroxid

Purity: 76.5% min CAS No.: 21645-51-2 Molecular Formula: AlH3O3 Formula Weight: 78.00 Physical Form: Granular Appearance: White Solubility: Practically insoluble in water. Soluble in alkaline aqueous solution, HCl, H2SO4 solution Merck Reference: 14,342 MDL No.: MFCD00003420


As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics

Formel: Al(OH)₃
Schmelzpunkt: 300 °C
Dichte: 2,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003420
CAS-Nummer: 21645-51-2
EINECS: 244-492-7
Merck Index: 14,342

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR