Cadmium(II)sulfat 8/3-Hydrat 99,0-102,0% (durch EDTA Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Cadmium sulphate, 8/3-hydrate, Cadmium sulphate octahydrate
1243-04EA
156.74
EUR
1243-04
Cadmium(II)sulfat 8/3-Hydrat 99,0-102,0% (durch EDTA Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Cadmium(II)sulfat 8/3-Hydrat
Formel:
3CdSO₄·8H₂O Schmelzpunkt: 41 °C Dichte: 3,08 kg/l |
MDL:
MFCD02253000 CAS-Nummer: 7790-84-3 EINECS: 233-331-6 UN: 2570 ADR: 6.1,III Merck Index: 13,01627 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 99.0 - 102.0 % |
Assay (CdSO₄.8/3 H₂O) (by EDTA titrn) | 99.0 - 102.0 % |
Insoluble Matter | ≤0.005 % |
Chloride (Cl) | ≤0.001 % |
Nitrate and Nitrite (as NO₃) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.002 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Lead (Pb) | ≤0.001 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Zinc (Zn) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...