Gallocyanine 90% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
124508-10G
124508-10GEA
94.4
EUR
124508-10G
Gallocyanine 90% (Farbstoffgehalt), Sigma-Aldrich®
Gallocyanine
Gallocyanine, Dye content 90 % (7-Dimethylamino-4-hydroxy-3-oxo-phenoxazine-1-carboxylic acid)
Formel:
C₁₅H₁₃ClN₂O₅ Molecular Weight: 336,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011926 CAS-Nummer: 1562-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...