Blei(IV)oxid 97,0 ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bleidioxid
12483.22
12483.36
12483.22EA
73
EUR
12483.22
12483.36
Blei(IV)oxid 97,0 ACS
Blei(IV)oxid
Purity: 97.0% min
CAS No.: 1309-60-0
Molecular Formula: PbO2
Formula Weight: 239.19
Physical Form: Powder
Density: 9.38
Melting Point: 290° dec.
Solubility: Insoluble in water. Soluble in HCl (with evolution of chlorine), oxalic acid, HNO3 in presence of H2O2, alkali iodide solutions, hot alkali caustic solutions
Merck Reference: 14,5407
UN No.: 1872
MDL No.: MFCD00011165
In battery electrodes, manufacturing of pigments and dyes, as analytical lab reagent
Formel:
O₂Pb Molecular Weight: 239.2 g/mol Schmelzpunkt: 290 °C (Zers.) Dichte: 9.38 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011165 CAS-Nummer: 1309-60-0 EINECS: 215-174-5 UN: 1872 ADR: 5.1 III Merck Index: 14,5407 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...