Iron(II) chloride hydrate ≥99%, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 23838-02-0
Molecular Formula: Cl2Fe.xH2O
Formula Weight: 126.75(anhy)
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Green-yellow
Odour: Odourless
Solubility: Soluble in water, alcohol and acetone. Insoluble in ether
UN No.: UN3260
MDL No.: MFCD00149711
Applications: Reducing agent in metallurgy, in pharmaceutical preparations, as mordant in dyeing and in sewage treatment
Formel:
Cl₂Fe·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149711 CAS-Nummer: 23838-02-0 EINECS: 231-843-4 UN: 3260 ADR: 8,III Merck Index: 14,4043 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...