Strontiumchlorid Hexahydrat, tech.
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 10025-70-4
Molecular Formula: SrCl2.6H2O
Formula Weight: 266.62 (158.53anhy)
Physical Form: Crystalline
Density: 1.93
Refractive Index: 1.650
Sensitivity: Hygroscopic
Appearance: White to off-white
Solubility: Soluble in water, alcohol
Merck Reference: 14,8840
MDL No.: MFCD00149865
Formel:
SrCl₂·6H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 61 °C Dichte: 1,93 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149865 CAS-Nummer: 10025-70-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...