Eisen(III)chlorid Hexahydrat 97,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Eisentrichlorid Hexahydrat
12497.14
12497.30
12497.A1
12497.A7
12497.14EA
31.7
EUR
12497.14
12497.30
12497.A1
12497.A7
Eisen(III)chlorid Hexahydrat 97,0-102,0% ACS
Eisen(III)chlorid Hexahydrat
Purity: 97.0-102.0%
CAS No.: 10025-77-1
Molecular Formula: Cl3Fe.6H2O
Formula Weight: 270.30 (162.21anhy)
Physical Form: Lump
Density: 1.820
Appearance: Yellow-brown
Odour: Odourless
pH: 1.8
Melting Point: 37°
Boiling Point: 280-285°
Solubility: Freely soluble in water, alcohol, ether, acetone.
Merck Reference: 14,4019
UN No.: 3260
MDL No.: MFCD00149712
Formel:
FeCl₃·6H₂O Siedepunkt: 280…285 °C Schmelzpunkt: 37 °C Dichte: 1,820 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149712 CAS-Nummer: 10025-77-1 EINECS: 231-729-4 UN: 3260 ADR: 8,III Merck Index: 14,4019 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...