L-Alanin, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
H-L-Ala-OH, (S)-(+)-2-Aminopropionic acid, L(+)-2-Aminopropionic acid, A, (S)-(+)-Alanin, α-Alanin, H-Ala-OH, (S)-(+)-2-Aminopropionsäure, L(+)-Alanin, (S)-(+)-2-Aminopropansäure, L-α-Alanin
1250-100
1250-100EA
107
EUR
1250-100
L-Alanin, Millipore®
L-Alanin
Formel:
C₃H₇NO₂ Molecular Weight: 89.09 g/mol Siedepunkt: 189 Schmelzpunkt: 79 Dichte: 1.4 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064410 CAS-Nummer: 56-41-7 EINECS: 200-273-8 Merck Index: 13,00203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...