Aluminiumoxid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Aluminium(III) oxide, Alumina C, Aluminum oxide

12553.36 12553.A3
12553.36EA 48.1 EUR
12553.36 12553.A3
Aluminiumoxid ≥99% (Metall-Basis)
Aluminium oxide

Purity: 99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: Fine Powder Density: 3.965 Refractive Index: 1.768 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424


Catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fibers

Formel: Al₂O₃
Molecular Weight: 101.96 g/mol
Siedepunkt: 2980 °C
Schmelzpunkt: 2045
Dichte: 3.965
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR