Dioctylphenylphosphonat, Supelco®
Lieferant: Merck
Synonyme:
Di-n-octyl phenylphosphonate
M-Clarity™ Qualitätsstufe = MQ100
12584-10ML
12584-10MLEA
476
EUR
12584-10ML
Dioctylphenylphosphonat, Supelco®
Dioctylphenylphosphonat
Formel:
C₂₂H₃₉O₃P Molecular Weight: 382,53 g/mol |
MDL:
MFCD00009561 CAS-Nummer: 1754-47-8 EINECS: 217-142-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...