Natriumselenat, anhydrous 99,8 Metals basis

Lieferant: Thermo Fisher Scientific
Gefahr

12613.18 12613.06 12613.30 12613.A1
12613.18EA 65.7 EUR
12613.18 12613.06 12613.30 12613.A1
Natriumselenat, anhydrous 99,8 Metals basis
Sodium selenate

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.


Stable under alkaline and oxidizing conditions.


As a reagent

Formel: Na₂O₄Se
Molecular Weight: 188.94 g/mol
Schmelzpunkt: 32 °C trans.
Dichte: 3.213
Lagertemperatur: Raumtemperatur
MDL: MFCD00003490
CAS-Nummer: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1 I
Merck Index: 14,8671

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR