Ruthenium(III) nitrosylnitrate-Lösung (1,5% Ru) (w/v)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12630.AD
12630.AK
12630.AU
12630.ADEA
116
EUR
12630.AD
12630.AK
12630.AU
Ruthenium(III) nitrosylnitrate-Lösung (1,5% Ru) (w/v)
CAS No.: 34513-98-9
Molecular Formula: N4O10Ru
Formula Weight: 317.09
Physical Form: Liquid
Density: 1.07
Appearance: Red-brown
Odour: Acidic
UN No.: UN3093
MDL No.: MFCD00016313
Formel:
N₄O₁₀Ru Dichte: 1,07 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016313 CAS-Nummer: 34513-98-9 EINECS: 252-068-8 UN: 3093 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...