Kaliumosmat(VI) Dihydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12647.01
12647.03
12647.01EA
66.4
EUR
12647.01
12647.03
Kaliumosmat(VI) Dihydrat ≥98%
Kaliumosmat(VI) Dihydrat
Purity: 98+%
CAS No.: 10022-66-9
Molecular Formula: K2OsO4.2H2O
Formula Weight: 368.43 (332.40anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Purple
Solubility: Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol, ether
UN No.: 3288
MDL No.: MFCD00149919
Formel:
K₂O₄Os·2H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149919 CAS-Nummer: 10022-66-9 EINECS: 243-247-1 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...