Ethyl Acrylate and Methyl Methacrylate Copolymer Dispersion, Reference Standard
Lieferant: USP
1265424
1265424.EA
452
EUR
1265424.
Ethyl Acrylate and Methyl Methacrylate Copolymer Dispersion, Reference Standard
Standards
Organische Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Standards: US Pharmacopea Reference Standard