Aldehyde dehydrogenase
Lieferant: Merck Millipore Calbiochem
126925-100
126925-100EA
260
EUR
126925-100
Aldehyde dehydrogenase
Enzyme
Aldehyde dehydrogenase is a native aldehyde dehydrogenase from yeast that catalyses the oxidation of acetaldehyde to acetic acid. Used for the determination of acetaldehyde.
- Form: White lyophilised solid
- Specific activity: ≥20 units/mg protein
- Storage temperature: 2 to 8 °C
One unit is defined as the amount of enzyme that will oxidise 1,0 µmol of acetaldehyde to acetic acid per min at 25 °C, pH 8,0.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...