Zink 99,9998 Metals basis, rods, Ø 10 mm (0.39 in)
Lieferant: Thermo Fisher Scientific
12718.DT
12718.DTEA
597
EUR
12718.DT
Zink 99,9998 Metals basis, rods, Ø 10 mm (0.39 in)
Zinc
Purity: 99.9998%
CAS No.: 7440-66-6
Physical Form: ≈5.60g/cm
Odour: Odourless
MDL No.: MFCD00011291
Formel:
Zn Molecular Weight: 65.39 g/mol Siedepunkt: 908 Schmelzpunkt: 420 Dichte: 7.14 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011291 CAS-Nummer: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...