Dichloro(1,5-cyclooctadiene)platinum(II) (51,6 - 52,6% Pt)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12739.01
12739.03
12739.06
12739.01EA
79.5
EUR
12739.01
12739.03
12739.06
Dichloro(1,5-cyclooctadiene)platinum(II) (51,6 - 52,6% Pt)
Dichloro(1,5-cyclooctadiene)platinum(II)
CAS No.: 12080-32-9
Molecular Formula: C8H12Cl2Pt
Formula Weight: 374.18
Physical Form: Solid
Sensitivity: Hygroscopic
Melting Point: 250° dec.
Solubility: Soluble in dichloromethane
MDL No.: MFCD00012413
Formel:
C₈H₁₂Cl₂Pt Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012413 CAS-Nummer: 12080-32-9 EINECS: 235-144-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...