Methylsuccinic anhydride 97%

Lieferant: Thermo Fisher Scientific
Achtung

127750050 127750250
127750050.EA 69.1 EUR
127750050. 127750250.
Methylsuccinic anhydride 97%
Methylsuccinic anhydride
Formel: C₅H₆O₃
Siedepunkt: 238…240°C (760mmHg)
Dichte: 1,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00005527
CAS-Nummer: 4100-80-5
EINECS: 223-870-5

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow low melting solid
Infrared spectrum Conforms
Titration by coupling 96.0 to 104.0 %
Melting point 33°C to 35°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR