Diarsentriselenid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.999%
CAS No.: 1303-36-2
Molecular Formula: As2Se3
Formula Weight: 386.72
Physical Form: 1-6mm Fused Pieces
Density: 4.75
Melting Point: 260°
Merck Reference: 14,805
UN No.: 3077
MDL No.: MFCD00014166
For vacuum deposition
Formel:
As₂Se₃ Schmelzpunkt: 260 °C Dichte: 4,75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014166 CAS-Nummer: 1303-36-2 EINECS: 215-119-5 UN: 3077 ADR: 9,III Merck Index: 14,805 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...