Calciumcarbonat ≥99,0% (Trockenmasse, T), Pulver, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor

Synonyme: Marmor

1288-REA 0 EUR
1288-R
Calciumcarbonat ≥99,0% (Trockenmasse, T), Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Calciumcarbonat

Powder. Lot analysis on label.


  • Lot analysis on label.

Formel: CaCO₃
Molecular Weight: 100,09 g/mol
Schmelzpunkt: 825 °C
Dichte: 2,71 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010906
CAS-Nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (CaCO₃) (dried basis) ≥99.0 %
Insoluble in Dilute HCl ≤0.01 %
Chloride (Cl) ≤0.001 %
Fluoride (F) ≤0.0015 %
Sulfate (SO₄) ≤0.01 %
Ammonium (NH₄) ≤0.003 %
Barium (Ba) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.003 %
Magnesium (Mg) ≤1.0 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.1 %
Strontium (Sr) ≤0.1 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR