Cäsiumcarbonat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12887.06
12887.18
12887.30
12887.A1
12887.06EA
25.8
EUR
12887.06
12887.18
12887.30
12887.A1
Cäsiumcarbonat ≥99% (Metall-Basis)
Cesium carbonate
Purity: 99%
CAS No.: 534-17-8
Molecular Formula: Cs2CO3
Formula Weight: 325.82
Physical Form: Powder/Granules
Density: 4.072
Appearance: White
Odour: Odourless
pH: 10-12
Melting Point: 610° dec.
Solubility: Very soluble in water and alcohol. Soluble in ether
Beilstein Reference: 4546405
Merck Reference: 14,2010
MDL No.: MFCD00010957
Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes
Formel:
Cs₂CO₃ Molecular Weight: 325.82 g/mol Schmelzpunkt: 610 °C (Zers.) Dichte: 4.072 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010957 CAS-Nummer: 534-17-8 EINECS: 208-591-9 Merck Index: 14,2010 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...