Bucket, Rectangular
Lieferant: FHP VILEDA SCS
120942 131662
129-0458EA 26.1 EUR
129-0458 129-0454
Bucket, Rectangular
Eimer
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
  • Liquid measurement lines molded inside of bucket
  • Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
  • Autoclavable
Highest performance for the best contamination control.