Sweeper Pan
Lieferant: FHP VILEDA SCS
119914
129-0460EA 59.7 EUR
129-0460
Sweeper Pan
Kehrschaufeln
The Sweeper and Pan is the perfect system for ergonomic and efficient dirt and liquid removal.
  • Flexible joint allows the user to reach into the most narrow spaces
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the cleanroom
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
The system is hygienic because of the washable and non-corroding material used in the construction. Overlapping rubber at the edges of the squeegee protect walls and furniture and the ergonomic grip reduces worker fatigue. Perfect for dry or damp conditions on smooth floors.
Bestellinformation: Sweeper and pan have to be ordered separately.